Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743174.4 | XP_002743220.1 | 1 - 2 | 560 | 82 | |
| XM_009000848.2 | XP_008999096.1 | 1 - 2 | 237 | 82 | ||
More Information
Set Membership: |
|

TMEM38B
Genomic Map