Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994357.1 | XP_008992605.1 | 2 - 3 | 409 | 69 | |
| XM_017971051.1 | XP_017826540.1 | 2 - 3 | 409 | 69 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map