Assay Details
| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964792.1 | XP_017820281.1 | 1 | 5413 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC15| Gene Symbol: LRRC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964792.1 | XP_017820281.1 | 1 | 5413 | 110 | |
Set Membership: |
|