Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745667.2 | XP_002745713.1 | 3 - 4 | 759 | 74 | |
| XM_008993090.2 | XP_008991338.1 | 2 - 3 | 320 | 74 | ||
| XM_008993091.1 | XP_008991339.1 | 3 - 4 | 730 | 74 | ||
| XM_017970465.1 | XP_017825954.1 | 2 - 3 | 223 | 74 | ||
More Information
Set Membership: |
|

ENOPH1
Genomic Map