Assay Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758412.4 | XP_002758458.1 | 6 - 7 | 1542 | 59 | |
| XM_008981774.2 | XP_008980022.1 | 6 - 7 | 1542 | 59 | ||
| XM_008981775.2 | XP_008980023.1 | 6 - 7 | 1542 | 59 | ||
| XM_008981776.2 | XP_008980024.1 | 6 - 7 | 1537 | 59 | ||
| XM_008981778.2 | XP_008980026.1 | 4 - 5 | 519 | 59 | ||
| XM_008981779.2 | XP_008980027.1 | 4 - 5 | 792 | 59 | ||
| XM_017964942.1 | XP_017820431.1 | 6 - 7 | 1538 | 59 | ||
| XM_017964943.1 | XP_017820432.1 | 5 - 6 | 667 | 59 | ||
| XM_017964944.1 | XP_017820433.1 | 6 - 7 | 1542 | 59 | ||
More Information
Set Membership: |
|

PPM1M
Genomic Map