Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257217.1 | NP_001244146.1 | 3 - 4 | 645 | 75 | |
| GenBank mRNA | EF534206.1 | - | 3 - 4 | 645 | 75 | |
More Information
Set Membership: |
|

CD2
Genomic Map