Assay Details
| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752356.4 | XP_002752402.1 | 4 - 5 | 1443 | 91 | |
| XM_009003608.2 | XP_009001856.1 | 4 - 5 | 1362 | 91 | ||
| XM_009003610.2 | XP_009001858.1 | 4 - 5 | 620 | 91 | ||
More Information
Set Membership: |
|

PUS7L
Genomic Map