Assay Details
Gene Symbol: HEXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008997972.2 | XP_008996220.1 | 8 - 9 | 1040 | 65 | |
XM_017972959.1 | XP_017828448.1 | 7 - 8 | 1396 | 65 | ||
XM_017972960.1 | XP_017828449.1 | 6 - 7 | 1231 | 65 | ||
XM_017972961.1 | XP_017828450.1 | 7 - 8 | 1406 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
