Assay Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754263.4 | XP_002754309.1 | 2 - 3 | 373 | 67 | |
| XM_017977320.1 | XP_017832809.1 | 2 - 3 | 283 | 67 | ||
More Information
Set Membership: |
|

GSKIP
Genomic Map