Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757848.3 | XP_002757894.1 | 5 - 6 | 1007 | 73 | |
| XM_008980854.1 | XP_008979102.1 | 5 - 6 | 1629 | 73 | ||
| XM_008980855.2 | XP_008979103.1 | 4 - 5 | 1351 | 73 | ||
More Information
Set Membership: |
|

QPCT
Genomic Map