Assay Details
| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747854.2 | XP_002747900.1 | 5 - 6 | 2309 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM83G| Gene Symbol: FAM83G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747854.2 | XP_002747900.1 | 5 - 6 | 2309 | 55 | |
Set Membership: |
|