Assay Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744429.4 | XP_002744475.1 | 5 - 6 | 383 | 90 | |
| XM_008988695.2 | XP_008986943.1 | 5 - 6 | 401 | 90 | ||
| XM_008988709.1 | XP_008986957.1 | 4 - 5 | 240 | 90 | ||
More Information
Set Membership: |
|

MRPL22
Genomic Map