Assay Details
| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001197.2 | XP_008999445.1 | 6 - 7 | 790 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGBL4| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001197.2 | XP_008999445.1 | 6 - 7 | 790 | 60 | |
Set Membership: |
|