Assay Details
| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001197.2 | XP_008999445.1 | 9 - 10 | 1111 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGBL4| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001197.2 | XP_008999445.1 | 9 - 10 | 1111 | 76 | |
Set Membership: |
|