Assay Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968398.1 | XP_017823887.1 | 6 - 7 | 853 | 65 | |
| XM_017968399.1 | XP_017823888.1 | 6 - 7 | 853 | 65 | ||
| XM_017968400.1 | XP_017823889.1 | 6 - 7 | 853 | 65 | ||
More Information
Set Membership: |
|

GYG2
Genomic Map