Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983430.2 | XP_008981678.1 | 2 - 3 | 1381 | 71 | |
| XM_008983431.2 | XP_008981679.1 | 2 - 3 | 1381 | 71 | ||
More Information
Set Membership: |
|

ERICH5
Genomic Map