Assay Details
| Gene Symbol: GPR39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749508.3 | XP_002749554.1 | 1 - 2 | 1312 | 58 | |
| XM_008998998.2 | XP_008997246.1 | 2 - 3 | 1102 | 58 | ||
More Information
Set Membership: |
|

GPR39
Genomic Map