Assay Details
| Gene Symbol: RABGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743900.4 | XP_002743946.2 | 8 - 9 | 2301 | 90 | |
| XM_008981222.1 | XP_008979470.1 | 9 - 10 | 1311 | 90 | ||
| XM_008981227.1 | XP_008979475.1 | 8 - 9 | 1252 | 90 | ||
| XM_008981234.2 | XP_008979482.1 | 9 - 10 | 1274 | 90 | ||
| XM_008981236.1 | XP_008979484.1 | 8 - 9 | 1239 | 90 | ||
| XM_008981243.1 | XP_008979491.1 | 8 - 9 | 1249 | 90 | ||
| XM_017965875.1 | XP_017821364.1 | 8 - 9 | 2305 | 90 | ||
| XM_017965887.1 | XP_017821376.1 | 8 - 9 | 2302 | 90 | ||
| XM_017965894.1 | XP_017821383.1 | 5 - 6 | 733 | 90 | ||
More Information
Set Membership: |
|

RABGEF1
Genomic Map