Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994920.2 | XP_008993168.1 | 3 - 4 | 623 | 101 | |
| XM_008994921.2 | XP_008993169.1 | 3 - 4 | 422 | 101 | ||
| XM_008994922.2 | XP_008993170.1 | 3 - 4 | 419 | 101 | ||
| XM_008994923.2 | XP_008993171.1 | 3 - 4 | 620 | 101 | ||
| XM_008994924.2 | XP_008993172.1 | 3 - 4 | 567 | 101 | ||
| XM_008994925.2 | XP_008993173.1 | 3 - 4 | 366 | 101 | ||
| XM_008994926.2 | XP_008993174.1 | 3 - 4 | 363 | 101 | ||
| XM_017971329.1 | XP_017826818.1 | 3 - 4 | 333 | 101 | ||
| XM_017971330.1 | XP_017826819.1 | 3 - 4 | 534 | 101 | ||
More Information
Set Membership: |
|

FAM229B
Genomic Map