Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761915.2 | XP_002761961.1 | 7 - 8 | 988 | 66 | |
| XM_008987533.1 | XP_008985781.1 | 6 - 7 | 1085 | 66 | ||
More Information
Set Membership: |
|

TMEM59L
Genomic Map