Assay Details
| Gene Symbol: CYP2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754993.3 | XP_002755039.1 | 2 - 3 | 413 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2R1| Gene Symbol: CYP2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754993.3 | XP_002755039.1 | 2 - 3 | 413 | 73 | |
Set Membership: |
|