Assay Details
| Gene Symbol: ADM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735635.3 | XP_003735683.2 | 1 | 595 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003735635.3 transcripts.
Set Membership: |
|

ADM5
Genomic Map