Assay Details
| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743816.4 | XP_002743862.2 | 4 - 5 | 710 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLDIP3| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743816.4 | XP_002743862.2 | 4 - 5 | 710 | 61 | |
Set Membership: |
|