Assay Details
| Gene Symbol: WIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975680.1 | XP_017831169.1 | 4 - 5 | 1238 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WIPF3| Gene Symbol: WIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975680.1 | XP_017831169.1 | 4 - 5 | 1238 | 69 | |
Set Membership: |
|