Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745875.3 | XP_002745921.1 | 3 - 4 | 1246 | 75 | |
| XM_008993404.2 | XP_008991652.1 | 3 - 4 | 404 | 75 | ||
| XM_017970584.1 | XP_017826073.1 | 3 - 4 | 404 | 75 | ||
More Information
Set Membership: |
|

GABRA4
Genomic Map