Assay Details
| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980728.1 | XP_008978976.1 | 745 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPCAM| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980728.1 | XP_008978976.1 | 745 | 85 | ||