Assay Details
| Gene Symbol: MDH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999328.2 | XP_008997576.1 | 12 | 2126 | 98 | |
| XM_008999329.2 | XP_008997577.1 | 11 | 2059 | 98 | ||
| XM_017973670.1 | XP_017829159.1 | 11 | 2051 | 98 | ||
More Information
Set Membership: |
|

MDH1B
Genomic Map