Assay Details
| Gene Symbol: LRRC47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750216.3 | XP_002750262.1 | 5 - 6 | 1460 | 78 | |
| XM_009000302.2 | XP_008998550.1 | 5 - 6 | 1460 | 78 | ||
More Information
Set Membership: |
|

LRRC47
Genomic Map