Assay Details
| Gene Symbol: GFY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762369.4 | XP_002762415.2 | 2 | 860 | 72 | |
| XM_017967886.1 | XP_017823375.1 | 3 | 979 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFY| Gene Symbol: GFY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762369.4 | XP_002762415.2 | 2 | 860 | 72 | |
| XM_017967886.1 | XP_017823375.1 | 3 | 979 | 72 | ||
Set Membership: |
|