Assay Details
| Gene Symbol: GUK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985667.2 | XP_008983915.2 | 7 - 8 | 610 | 69 | |
| XM_017966665.1 | XP_017822154.1 | 6 - 7 | 609 | 69 | ||
| XM_017966666.1 | XP_017822155.1 | 6 - 7 | 678 | 69 | ||
| XM_017966667.1 | XP_017822156.1 | 5 - 6 | 631 | 69 | ||
| XM_017966669.1 | XP_017822158.1 | 5 - 6 | 532 | 69 | ||
| XM_017966670.1 | XP_017822159.1 | 8 - 9 | 708 | 69 | ||
| XM_017966671.1 | XP_017822160.1 | 5 - 6 | 449 | 69 | ||
More Information
Set Membership: |
|

GUK1
Genomic Map