Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755138.4 | XP_002755184.1 | 4 - 5 | 508 | 98 | |
| XM_009007869.2 | XP_009006117.1 | 4 - 5 | 454 | 98 | ||
| XM_017977978.1 | XP_017833467.1 | 4 - 5 | 419 | 98 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map