Assay Details
| Gene Symbol: PRRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008990040.2 | XP_008988288.1 | 3 - 4 | 315 | 88 | |
| XM_017968690.1 | XP_017824179.1 | 3 - 4 | 328 | 88 | ||
| XM_017968691.1 | XP_017824180.1 | 3 - 4 | 291 | 88 | ||
| XM_017968692.1 | XP_017824181.1 | 3 - 4 | 303 | 88 | ||
More Information
Set Membership: |
|

PRRG3
Genomic Map