Assay Details
| Gene Symbol: MINDY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759896.3 | XP_002759942.1 | 6 - 7 | 1086 | 64 | |
| XM_008984310.1 | XP_008982558.1 | 6 - 7 | 1865 | 64 | ||
| XM_008984311.2 | XP_008982559.1 | 6 - 7 | 1224 | 64 | ||
| XM_008984312.1 | XP_008982560.1 | 6 - 7 | 1862 | 64 | ||
| XM_008984313.1 | XP_008982561.1 | 6 - 7 | 1865 | 64 | ||
| XM_008984314.1 | XP_008982562.1 | 6 - 7 | 1865 | 64 | ||
| XM_008984315.1 | XP_008982563.1 | 6 - 7 | 1865 | 64 | ||
| XM_008984316.1 | XP_008982564.1 | 5 - 6 | 1703 | 64 | ||
| XM_017966251.1 | XP_017821740.1 | 5 - 6 | 1846 | 64 | ||
More Information
Set Membership: |
|

MINDY1
Genomic Map