Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755690.3 | XP_002755736.1 | 3 - 4 | 864 | 68 | |
| XM_002755691.3 | XP_002755737.1 | 3 - 4 | 945 | 68 | ||
| XM_009008780.2 | XP_009007028.1 | 3 - 4 | 870 | 68 | ||
More Information
Set Membership: |
|

HRAS
Genomic Map