Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746080.3 | XP_002746126.1 | 1 - 2 | 1895 | 82 | |
| XM_017970809.1 | XP_017826298.1 | 2 - 3 | 1855 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED9| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746080.3 | XP_002746126.1 | 1 - 2 | 1895 | 82 | |
| XM_017970809.1 | XP_017826298.1 | 2 - 3 | 1855 | 82 | ||