Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745107.4 | XP_002745153.1 | 2 - 3 | 335 | 97 | |
| XM_008992233.2 | XP_008990481.1 | 2 - 3 | 269 | 97 | ||
More Information
Set Membership: |
|

OTULINL
Genomic Map