Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745107.4 | XP_002745153.1 | 6 - 7 | 740 | 99 | |
| XM_008992233.2 | XP_008990481.1 | 6 - 7 | 674 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTULINL| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745107.4 | XP_002745153.1 | 6 - 7 | 740 | 99 | |
| XM_008992233.2 | XP_008990481.1 | 6 - 7 | 674 | 99 | ||