Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745107.4 | XP_002745153.1 | 7 - 8 | 1006 | 109 | |
| XM_008992233.2 | XP_008990481.1 | 7 - 8 | 940 | 109 | ||
More Information
Set Membership: |
|

OTULINL
Genomic Map