Assay Details
| Gene Symbol: PGAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976521.1 | XP_017832010.1 | 2 - 3 | 459 | 61 | |
| XM_017976522.1 | XP_017832011.1 | 2 - 3 | 459 | 61 | ||
| XM_017976523.1 | XP_017832012.1 | 2 - 3 | 459 | 61 | ||
| XM_017976524.1 | XP_017832013.1 | 459 | 61 | |||
More Information
Set Membership: |
|

PGAM5
Genomic Map