Assay Details
| Gene Symbol: LOC100397870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972421.1 | XP_017827910.1 | 12 - 13 | 1392 | 65 | |
| XM_017972422.1 | XP_017827911.1 | 11 - 12 | 1275 | 65 | ||
More Information
Set Membership: |
|

LOC100397870
Genomic Map