Assay Details
| Gene Symbol: LOC100397870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972421.1 | XP_017827910.1 | 5 - 6 | 803 | 54 | |
| XM_017972422.1 | XP_017827911.1 | 5 - 6 | 803 | 54 | ||
More Information
Set Membership: |
|

LOC100397870
Genomic Map