Assay Details
| Gene Symbol: LOC100397870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972421.1 | XP_017827910.1 | 9 - 10 | 1040 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100397870| Gene Symbol: LOC100397870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972421.1 | XP_017827910.1 | 9 - 10 | 1040 | 110 | |
Set Membership: |
|