Assay Details
| Gene Symbol: PAICS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993307.2 | XP_008991555.1 | 8 - 9 | 1641 | 71 | |
| GenBank mRNA | EF214946.1 | - | 3 - 4 | 398 | 71 | |
More Information
Set Membership: |
|

PAICS
Genomic Map