Assay Details
| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988105.2 | XP_008986353.1 | 10 - 11 | 1841 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSK3A| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988105.2 | XP_008986353.1 | 10 - 11 | 1841 | 68 | |
Set Membership: |
|