Assay Details
| Gene Symbol: C5H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748380.3 | XP_002748426.1 | 9 - 10 | 1039 | 82 | |
| XM_008997106.2 | XP_008995354.1 | 10 - 11 | 1004 | 82 | ||
| XM_017972539.1 | XP_017828028.1 | 9 - 10 | 918 | 82 | ||
More Information
Set Membership: |
|

C5H17orf75
Genomic Map