Assay Details
| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005811.2 | XP_009004059.2 | 7 - 8 | 1524 | 100 | |
| XM_017977018.1 | XP_017832507.1 | 7 - 8 | 1596 | 100 | ||
| XM_017977019.1 | XP_017832508.1 | 7 - 8 | 1606 | 100 | ||
| XM_017977020.1 | XP_017832509.1 | 6 - 7 | 1232 | 100 | ||
More Information
Set Membership: |
|

IRF9
Genomic Map