Assay Details
| Gene Symbol: OTUD6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759060.3 | XP_002759106.3 | 4 - 5 | 826 | 109 | |
| XM_017965442.1 | XP_017820931.1 | 4 - 5 | 821 | 109 | ||
| XM_017965443.1 | XP_017820932.1 | 2 - 3 | 1096 | 109 | ||
More Information
Set Membership: |
|

OTUD6B
Genomic Map