Assay Details
| Gene Symbol: MRPS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748394.4 | XP_002748440.1 | 2 - 3 | 284 | 69 | |
| XM_008997311.2 | XP_008995559.1 | 2 - 3 | 264 | 69 | ||
More Information
Set Membership: |
|

MRPS23
Genomic Map