Assay Details
| Gene Symbol: ACTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756377.4 | XP_002756423.1 | 6 - 7 | 912 | 65 | |
| XM_009009932.2 | XP_009008180.1 | 6 - 7 | 2704 | 65 | ||
More Information
Set Membership: |
|

ACTA2
Genomic Map