Assay Details
| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007201.2 | XP_009005449.1 | 4 - 5 | 715 | 67 | |
| XM_017978193.1 | XP_017833682.1 | 4 - 5 | 716 | 67 | ||
More Information
Set Membership: |
|

TMEM191C
Genomic Map